Would you like to log in again?
SV System Performance Check Mix (SPCC) (4 Components), Restek
Supplier: Restek
31689
75834-748EA 64.02 USD InStock
75834-748
SV System Performance Check Mix (SPCC) (4 Components), Restek
Standards Organic Standards
This SV system performance check mix consist of four components: 2,4-Dinitrophenol, Hexachlorocyclopentadiene, 4-Nitrophenol and N-Nitroso-di-n-propylamine.
Shelf life 6 to 24 months.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty