Would you like to log in again?
Method 525.2 Internal Standard Mix, Restek
Supplier: Restek
31825
75834-790EA 39.77 USD InStock
75834-790
Method 525.2 Internal Standard Mix, Restek
Standards Environmental Analysis Standards
This mix contains 3 components: acenaphthene-d10, chrysene-d12 and phenanthrene-d10.
Certifications: Certified reference materials (CRMs) manufactured and QC tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty