Would you like to log in again?
Semivolatiles MegaMix Standard, EPA Method 625, (54 Components), Restek
Supplier: Restek
MegaMix®
31829
75834-796EA 260.93 USD InStock
75834-796
Semivolatiles MegaMix Standard, EPA Method 625, (54 Components), Restek
Standards Environmental Analysis Standards
This contais 54 components, incorporates all phenols, for fast preparation and calibration, incorporates 'additional' extractable method 625 compounds. GC-MS method applicable to analysis of organic compounds in water and soil.
Some of the target compounds in method 625 are subject to thermal or chemical degradation and consequent unpredictable chromatographic performance. hexachlorocyclopentadiene can decompose at the GC inlet. N-nitrosodimethylamine is difficult to separate from the solvent; also suggests that the analyst follow method 607 to identify this analyte. Some phenolic compounds in the target list, including chlorinated, methylated, and nitro-substituted phenols, are susceptible to adsorption and/or thermal degradation at the GC inlet. In addition, peak tailing can be a problem with phenols.
Diphenylamine is included, rather than N-nitrosodiphenylamine, in the MegaMix formulation because N-nitrosodiphenylamine decomposes to diphenylamine at the GC inlet.
Additional compounds in method 625 (listed compound N-nitrosodiphenylamine decomposes to MegaMix component diphenylamine). The six other additional compounds are components in othe reference mixes used for method 625: benzidine is included is β-BHC, δ-BHC, endosulfan I, endosulfan II, endrin.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty