Would you like to log in again?
LC Multiresidue Pesticide Standard #9 (7 Components), Restek
Supplier: Restek
31980
75834-900EA 141.87 USD InStock
75834-900
LC Multiresidue Pesticide Standard #9 (7 Components), Restek
Standards Organic Standards
Accurately detect and quantify pesticides of global food safety concern in a wide range of fruits, vegetables, and other commodities by LC-MS/MS.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Carbamate/uron compounds
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimized multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Consists of 7 components, those are aminocarb, desmedipham, formetanate hcl, mexacarbate, monceren, phenmedipham and propamocarb free base.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: The complete 204-compound kit also available (cat. # 31971) that covers many LC-determined pesticides listed by government agencies, other ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 204 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty