Would you like to log in again?
GC Multiresidue Pesticide Standard #7, (10 components), Restek
Supplier: Restek
32569
75835-058EA 214.21 USD InStock
75835-058
GC Multiresidue Pesticide Standard #7, (10 components), Restek
Standards Organic Standards
The multiresidue accurately identify and quantify pesticide residues by gc-ms/ms in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea, and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Formulated and grouped for maximum long-term stability
The multiresidue pesticide standard consist of ten components acequinocyl, bromopropylate, carfentrazone ethyl, chlorobenzilate, chlorpropham, chlozolinate, DCPA methyl ester (chlorthal-dimethyl), fluazifop-p-butyl, metalaxyl and 2-phenylphenol.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Ordering information: * NOTE: When combining a large number of compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analyzing each mix separately to ensure accurate results for every compound.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty