Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
B/N Surrogate Mix (4/89 SOW), Restek
Supplier: Restek
33028 31062 31024-25 31024 31024-15 31062-15 31086
75835-104EA 272.57 USD InStock
75835-104 75834-608 RK31062 75835-620 RK31086 75834-598 RK31024
B/N Surrogate Mix (4/89 SOW), Restek
Standards Environmental Analysis Standards
Base/neutrals surrogate standards (3 components) are packaged in methylene chloride in an ampoule.
  • Convenient 1 mL 5 mL and 10 mL package sizes
  • Highest concentrations commercially available
  • High purity for consistent results
Mixture contains 2-Fluorobiphenyl, Nitrobenzene-d5, and p-Terphenyl-d14
Delivery information: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty