Would you like to log in again?
Acid Surrogate Mix (4/89 SOW), Restek
Supplier: Restek
31063-15 33029 31063 31087 31025 31025-15 31025-25
RK31063EA 60.14 USD InStock
RK31063 RK31087 75834-610 RK31025 75834-600 75835-108 75834-602
Acid Surrogate Mix (4/89 SOW), Restek
Standards Environmental Analysis Standards
Standards are packaged in methanol in an ampule.
  • Convenient 1 mL, 5 mL and 10 mL package sizes
  • Highest concentrations commercially available
  • Reduces laboratory cost per sample extract
  • Mixture contains 2-Fluorophenol, Phenol-d6, and 2,4,6-Tribromophenol
Each standard includes an ampule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty