Would you like to log in again?
CARB 1004 Aldehyde/Ketone-DNPH Calibration Standard (13 Components), Restek
Supplier: Restek
33093
75835-152EA 107.67 USD InStock
75835-152
CARB 1004 Aldehyde/Ketone-DNPH Calibration Standard (13 Components), Restek
Standards Organic Standards
The CARB 1004 aldehyde/ketone-DNPH calibration standard consists of 13 components, they are acetaldehyde-2,4-DNPH, acetone-2,4-DNPH, acrolein-2,4-DNPH, benzaldehyde-2,4-DNPH, n-butyraldehyde-2,4-DNPH, crotonaldehyde-2,4-DNPH, formaldehyde-2,4-DNPH, hexaldehyde-2,4-DNPH, methacrolein-2,4-DNPH, methyl ethyl ketone-2,4-DNPH, propionaldehyde-2,4-DNPH, m-tolualdehyde-2,4-DNPH, valeraldehyde-2,4-DNPH.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty