Would you like to log in again?
ISO-BB Column Test Mix, Restek
Supplier: Restek
35290
75835-392EA 42.68 USD InStock
75835-392
ISO-BB Column Test Mix, Restek
Standards Chromatography Standards
8 components
8 components
(C13) n-Tridecane (629-50-5)
(C14) n-Tetradecane (629-59-4)
(C15) n-Pentadecane (629-62-9)
(C16) n-Hexadecane (544-76-3)
Benzylamine (100-46-9)
Biphenyl (92-52-4)
4-Chlorophenol (106-48-9)
1-Undecanol (112-42-5)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty