Would you like to log in again?
Silylation Derivatization Reagents, Restek
Supplier: Restek
35602 35603
75835-398EA 154.23 USD InStock
75835-398 75835-396
Silylation Derivatization Reagents, Restek
Standards Organic Standards
Contains: 35600, 35601: MSTFA (N-methyl-N-trimethylsilytrifluoroacetamide) (24589-78-4); 35602, 35603: MSTFA w/1% TMCS (N-methyl-N-trimethylsilytrifluoroacetamide w/1% trimethylchlorosilane) (24589-78-4); 35604, 35605: BSTFA (N,O-bis[trimethylsilyl]trifluoroacetamide) (25561-30-2); 35606, 35607: BSTFA w/1% TMCS (N,O-bis[trimethylsilyltrifluoroacetamide] w/1% trimethylchlorosilane) (25561-30-2); 35608, 35610: MTBSTFA w/1% TBDMCS (N-methyl-N[tert-butyldimethylsilyl trifluoroacetamide] w/1% tert-butyldimethylchlorosilane) (77377-52-7); 35611, 35612: TMCS (trimethylchlorosilane) (75-77-4)
  • Replace active hydrogen, reducing polarity and making the compounds more volatile.
  • Increase stability of derivatives.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty