Would you like to log in again?
Residual Solvents Class 2 - Mix A (15 components), Restek
Supplier: Restek
36271
75835-454EA 68.87 USD InStock
75835-454
Residual Solvents Class 2 - Mix A (15 components), Restek
Standards Organic Standards
These mixtures reflect the changes made in USP30/NF25 effective July 1, 2008.
Contains: Acetonitrile, 2,05 mg/ml, chlorobenzene, 1,80 mg/ml, cyclohexane , 19,40 mg/ml, cis-1,2-dichloroethene, 4,70 mg/ml, trans-1,2-dichloroethene, 4,70 mg/ml, 1,4-dioxane, 1,90 mg/ml, ethylbenzene, 1,84 mg/ml, methanol, 15,00 mg/ml, methylcyclohexane, 5,90 mg/ml, methylene chloride (dichloromethane), 3,00 mg/ml, tetrahydrofuran, 3,45 mg/ml, toluene, 4,45 mg/ml, m-xylene, 6,51 mg/ml, o-xylene, 0,98 mg/ml, p-xylene, 1,52 mg/ml.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty