Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
EPA 524.3 Reference Standards (5 separate mixes, including a MegaMix standard), Restek
Supplier: Restek
MegaMix®
30014 30015 30017 30013 30016
75834-390EA 192.06 USD InStock
75834-390 75834-382 75834-384 75835-532 75834-386
EPA 524.3 Reference Standards (5 separate mixes, including a MegaMix standard), Restek
Standards Environmental Analysis Standards
In support of the U.S. Safe Drinking Water Act (SDWA), Restek offers a complete set of EPA 524.3 reference standards for the monitoring of purgeable organic compounds in drinking water—using as few as three ampuls! In addition, this collection of certified reference materials (CRMs) also covers the seven volatile organic compounds (VOCs) included in the Unregulated Contaminant Monitoring Rule 3 (UCMR3), which requires monitoring of all public drinking water systems with 10,000 or more customers.
  • Full 82-component EPA 524.3 list using as few as three ampuls—reduce prep time and chances for error or contamination
  • EPA 524.3 VOA MegaMix ampul includes oxygenates group—no need to order separately
  • Volatile gases prepared separately—replace shorter-life components without wasting money on full list
  • Two options for internal and surrogate standards—separate or combined mix
  • Certified reference materials (CRMs) manufactured and QC-tested in Restek’s ISO-accredited labs—satisfy your ISO requirements
  • Also ideal for surface water and groundwater testing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty