Would you like to log in again?
CA WIP VOA Standard (11 Components), Restek
Supplier: Restek
30236
75835-546EA 45.59 USD InStock
75835-546
CA WIP VOA Standard (11 Components), Restek
Standards Petroleum Industry Standards
For monitoring samples suspected of gasoline contamination requires laboratories to calibrate and report these compounds.
It consists of benzene, chlorobenzene, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, ethylbenzene, methyl tert-butyl ether, toluene, m-xylene, o-xylene, p-xylene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty