Would you like to log in again?
524 Calibration Mix #7A (5 Components) , Restek
Supplier: Restek
30300
75835-554EA 34.92 USD InStock
75835-554
524 Calibration Mix #7A (5 Components) , Restek
Standards Organic Standards
The calibration mix consists of five components they are acetone, 2-butanone, 2-hexanone, 4-methyl-2-pentanone and tetrahydrofuran.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty