Would you like to log in again?
Ketones Mix, 524.2 Rev. 4.1 (5 Components), Restek
Supplier: Restek
30602
75835-588EA 44.62 USD InStock
75835-588
Ketones Mix, 524.2 Rev. 4.1 (5 Components), Restek
Standards Organic Standards
The ketones mix consists of five components they are acetone, 2-butanone (MEK), 1,1-dichloro-2-propanone, 2-hexanone and 4-methyl-2-pentanone (MIBK).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty