Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
524.2 Surrogate Standard (2 Components), Restek
Supplier: Restek
30607
75835-596EA 36.86 USD InStock
75835-596
524.2 Surrogate Standard (2 Components), Restek
Standards Environmental Analysis Standards
It separate mixes for surrogates and internal standard.
  • Fortification solution combines surrogates and internal standard
  • Calibration mixes and all other quality control mixes also available
The standard consists of teo components, 1-bromo-4-fluorobenzene (BFB) and 1,2-dichlorobenzene-d4.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty