Would you like to log in again?
ASTM D3606 Calibration Standard #4 without Internal Standard (2 Components), Restek
Supplier: Restek
30650
75835-612EA 137.74 USD InStock
75835-612
ASTM D3606 Calibration Standard #4 without Internal Standard (2 Components), Restek
Standards Petroleum Industry Standards
The mix consists of benzene (0.67% vol/vol), toluene (5% vol/vol).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty