Would you like to log in again?
Alternate Boiling Point/Carbon Number Distribution Marker Stock Standard (9 Components), Restek
Supplier: Restek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
31639
75835-734EA 43.65 USD InStock
75835-734
Alternate Boiling Point/Carbon Number Distribution Marker Stock Standard (9 Components), Restek
Standards Petroleum Industry Standards
The mix consists of (C6) n-hexane, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C28) n-octacosane, (C35) n-pentatriacontane, (C36) n-hexatriacontane.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty