Would you like to log in again?
Haloacetic Acid Mix #2 (9 Components), Restek
Supplier: Restek
31646
75835-742EA 85.36 USD InStock
75835-742
Haloacetic Acid Mix #2 (9 Components), Restek
Standards Organic Standards
The mix consists of, bromochloroacetic acid (400 μg/ml), bromodichloroacetic acid (400 μg/ml), chlorodibromoacetic acid (1000 μg/ml), dibromoacetic acid (200 μg/ml), dichloroacetic acid (600 μg/ml), monobromoacetic acid (400 μg/ml), monochloroacetic acid (600 μg/ml), tribromoacetic acid (2000 μg/ml), trichloroacetic acid (200 μg/ml)
  • Prepared from purified neat reference materials for the highest quality
  • Lot-to-lot consistency ensures analytical reproducibility
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements. Meets Stage 1 requirements for US EPA Methods 552, 552.1 and 552.2 and meets requirements for Stage 2 DBPR.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty