Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Internal Standards and Surrogates for Method 552, Restek
Supplier: Restek
31653 31650 31648 31881 31655 31656
75835-744EA 40.74 USD InStock
75835-744 75834-836 75835-756 75835-754 75835-748 75835-758
Internal Standards and Surrogates for Method 552, Restek
Standards Environmental Analysis Standards
Internal Standard: cat.# 31648. Surrogate for Method 552, 552.1: cat.#'s 31650. Surrogates for Method 552.2: cat.#'s 31653, 31655, 31656. Surrogate for Method 552.3: cat.#'s 31881
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty