Would you like to log in again?
PAH Supplement Mix for Method 8100 (8 Components), Restek
Supplier: Restek
31857
75835-810EA 351.14 USD InStock
75835-810
PAH Supplement Mix for Method 8100 (8 Components), Restek
Standards Organic Standards
The mix consists of benzo(j)fluoranthene, dibenz(a,h)acridine, dibenz(a,j)acridine, 7H-dibenzo(c,g)carbazole, dibenzo(a,e)pyrene, dibenzo(a,h)pyrene, dibenzo(a,i)pyrene, 3-methylcholanthrene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty