Would you like to log in again?
515.4 Methylated Chlorinated Acids Mix (16 components), Restek
Supplier: Restek
32444
75835-876EA 155.2 USD InStock
75835-876
515.4 Methylated Chlorinated Acids Mix (16 components), Restek
Standards Organic Standards
Mix contains Acifluorfen methyl ester, Bentazon methyl ester, Chloramben methyl ester, Dalapon methyl ester, 2,4-D methyl ester, 2,4-DB methyl ester, DCPA methyl ester (Chlorthal-dimethyl), Dicamba methyl ester, 3,5-Dichlorobenzoic acid methyl ester, Dichlorprop methyl ester, Dinoseb methyl ether, Pentachloroanisole, Picloram methyl ester, Quinclorac methyl ester, 2,4,5-T methyl ester, 2,4,5-TP.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty