Would you like to log in again?
Inkjet Microscope Slides, Springside Scientific
Supplier: Springside Scientific
AB-45-IKJ-YL1B AHS45-IKJ-PK1B AB-45-IKJ-PK AHS45-IKJ-WH1B AHS45-IKJ-WH AB-45-IKJ-YL AHS45-IKJ-YL1B AB-45-IKJ-PK1B AB-45-IKJ-WH AHS45-IKJ-PK AB-45-IKJ-WH1B AHS45-IKJ-YL
75836-026CS 221.58 USD InStock
75836-026 75836-016 75836-018 75836-176 75836-022 75836-166 75836-024 75836-168 75836-172 75836-174 75836-020 75836-170
Inkjet Microscope Slides, Springside Scientific
Slides Microscope Slides
Inkjet Microscope Slides are manufactured from 3/4 white glass
All slides are standard size, with frosting optimized for Leica inkjet labelers, ensuring good barcode reads with each use. Slides are available charged or uncharged.
Ordering information: Slides are packed 72 per cardboard box. Each box is individually sealed. Each case contains 20 boxes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty