Would you like to log in again?
Superwhite Uncharged Microscope Slides, Springside Scientific
Supplier: Springside Scientific
HMW102N9P HMW102N9B HMW102F9B HMW102N9W-1B HMW102N9B-1B HMW102F9W-1B HMW102N9P-1B HMW102F9P HMW102N9W HMW102F9P-1B HMW102F9B-1B HMW102F9W
75836-198CS 19.78 USD InStock
75836-198 75836-056 75836-200 75836-058 75836-062 75836-194 75836-196 75836-064 75836-054 75836-190 75836-192 75836-060
Superwhite Uncharged Microscope Slides, Springside Scientific
Slides Microscope Slides
Microscope slides made of pure, super white glass which prevents auto-fluorescence
All slides are uncharged, and are uniformly 75x25x1mm. Colored frosting is optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Microscope slides made of pure, super white glass which prevents auto-fluorescence. All slides are uncharged, and are uniformly 75x25x1mm. Colored frosting is optimized for hot foil tape labelers such as the Slidemate and Primera.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty