Would you like to log in again?
Inkjet Superwhite Microscope Slides, Springside Scientific
Supplier: Springside Scientific
HMW202F9W-1B HMW202F9Y HMW232F9W-1B HMW232F9P-1B HMW202F9W HMW202F9P-1B HMW232F9Y HMW202F9P HMW232F9W HMW202F9Y-1B HMW232F9Y-1B HMW232F9P
75836-202CS 26.21 USD InStock
75836-202 75836-048 75836-204 75836-066 75836-046 75836-068 75836-074 75836-076 75836-206 75836-070 75836-072 75836-050
Inkjet Superwhite Microscope Slides, Springside Scientific
Slides Microscope Slides
Inkjet Superwhite Microscope Slides are made of pure, super white glass which prevents auto-fluorescence
They are uniformly 75x25x1mm. Colored frosting is optimized for Leica inkjet labelers, ensuring consistently good barcode reads.
Ordering information: Charged slides are packaged in plastic boxes. Uncharged slides are packaged in ardboard boxes. Each box contains 72 slides, along with fiber desiccant. Each box is individually sealed. Each case contains 20 boxes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty