Would you like to log in again?
Histology Control Slides, Peroidic Acid Schiff with Digestion (PASD), Springside Scientific
Supplier: Springside Scientific
CS-PSD20 CS-PSD100
75836-104CS 208.37 USD InStock
75836-104 75836-130
Histology Control Slides, Peroidic Acid Schiff with Digestion (PASD), Springside Scientific
Slides Microscopy Control Slides
100% Human Tissue.
Two stained controls in every box. Top-quality, pure white glass slides/coverglass.
Ordering information: Packaged 22 (20 controls plus 2 stained) or 100 (98 controls plus 2 stained) slides in a 25-postion or 100-position plastic slide-box.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty