Would you like to log in again?
Large Microscope Slides, Springside Scientific
Supplier: Springside Scientific
BSLD2X9-03 BSLD2X9-01
75836-214CS 77.81 USD InStock
75836-214 75836-212
Large Microscope Slides, Springside Scientific
Slides Microscope Slides
Large microscope slides are manufactured from 3/4 white glass
These oversized slides feature 90° corners, and measure 5 x 22.8 cm (2 x 9").
Ordering information: Large slides are packaged in a cardboard box, and are individually seperated by white paper.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty