Would you like to log in again?
Two-Circle Microscope Slides for Fluorescent Microscopy, Springside Scientific
Supplier: Springside Scientific
W1030C-C20
75836-216CS 377.31 USD InStock
75836-216
Two-Circle Microscope Slides for Fluorescent Microscopy, Springside Scientific
Slides Microscope Slides
Two-Circle Microscope Slides are manufactured from pure white glass, and feature two frosted circles designed for use in fluorescent antibody examination. Slides are standard size. The frosted cirscles measure 15 mm in diameter and are placed 11 mm apart. Two-circle slides are pre-cleaned, with 90° corners.
Slides are standard size. The frosted circles measure 15 mm in diameter and are placed 11 mm apart. Two-circle slides are pre-cleaned, with 90° corners.
Ordering information: Slides are packed 72 per cardboard box. Each box is individually sealed. Each case contains 20 boxes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty