Would you like to log in again?
Adhesive/Charged Microscope Slides, Springside Scientific
Supplier: Springside Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
AHS90-PK AHS45-PK AHS90-PK01-1B AHS45-WH01-1B AHS45-BL-1B AHS90-WH AHS90-BL01-1B AHS45-WH AHS90-WH-1B AHS45-BL AHS90-BL AHS45-PK-1B
75836-224CS 296.75 USD InStock
75836-224 75836-148 75836-226 75836-220 75836-144 75836-222 75836-002 75836-146 75836-150 75836-152 75836-142 75836-218
Adhesive/Charged Microscope Slides, Springside Scientific
Slides Adhesive Slides
Microscope slides are manufactured from 3/4 white glass and feature a positive charge for improved adhesion
All slides are standard size, with frosting optimized for hot foil tape labelers such as the Slidemate and Primera.
Ordering information: Slides are packed 72 per white cardboard box. Each box is individually sealed. Each case contains 20 boxes.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty