Would you like to log in again?
C. diff (Clostridium difficile) Banana Broth™, Hardy Diagnostics
Supplier: Hardy Diagnostics
K226
75836-228PK 138.33 USD InStock
75836-228
C. diff (Clostridium difficile) Banana Broth™, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
C. diff Banana Broth™ (Clostridium difficile Brucella Broth with Thioglycolic Acid and L-cystine or CDBB-TC) is recommended for the culture and recovery of Clostridium difficile spores and vegetative cells from environmental samples.
This product is not intended to be used for the diagnosis of human disease
C. diff Banana Broth™ is pre-reduced, ready to use, highly sensitive, and a specific detection method for Clostridium difficile. The high specificity and truly vivid color distinction makes environmental testing for Clostridium difficile extremely simple.
C. diff Banana Broth™ is a great tool to verify the efficacy of hospital cleaning procedures. Approximately 1.7 million hospitalized patients develop nosocomial infections anually while being treated for entirely unrelated conditions. Clostridium difficile has become increasingly common among nosocomial infections which makes screening for it increasingly important.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty