Would you like to log in again?
Orbital-Genie™ Mini-100 Orbital Shaker, Scientific Industries
Supplier: Scientific Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SI-M100
75836-230EA 1276.93 USD InStock
75836-230
Orbital-Genie™ Mini-100 Orbital Shaker, Scientific Industries
Shakers and Mixers Orbital Shakers
The Mini-100 Orbital-Genie with spill-proof edges is ideal for gentle shaking applications, with no additional accessories necessary.
  • Variable shaking speed
  • Customizable mixing action
  • Heavy-duty base
  • Holds oddly shaped containers
  • Ready-to-use
  • Two-year manufacturer's warranty
The Mini-100 Orbital-Genie features variable shaking speed, which allows you to create any mixing action from a smooth vortex to sloshing aeration or anything in between. The combination of weight and specially designed feet prevent walking even at highest speed. You can increase the capacity of the Mini-100 with an optional stacking tray, available separately. The Mini-100 is ideal for gentle shaking of odd shaped vessels and containers. The non-slip platform tray features edges that contain any spills. Shaker is ready to use as supplied.
Certifications: CE marked.
Accessories information: Optional stacking tray increases capacity.
Ordering information: Unit has a two-year manufacturer's warranty included.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty