Would you like to log in again?
Mini-300 Orbital Genie™, Scientific Industries
Supplier: Scientific Industries
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SI-M1603 SI-M1703 SI-M1600 SI-M1501 SI-M1502 SI-M1700 SI-M1702 SI-M1500 SI-M1602 SI-M1503 SI-M1701 SI-M1601
75836-246EA 1897.63 USD InStock
75836-246 75836-258 75836-248 75836-402 75836-254 75836-244 75836-256 75836-260 75836-250 75836-262 75836-252 75836-404
Mini-300 Orbital Genie™, Scientific Industries
Shakers and Mixers Orbital Shakers
Powerful high speed orbital shakers for use with virtually any size or shape container in three different configurations.
  • Super compact design
  • Powerful motor for shaking speed up to 300 RPM
  • Three models provides choice
  • Adhering mat model is ideal for open containers, bottles, flasks, or beakers
  • Ratcheting clamps model provides flexibility and stronghold of various size and shape containers
  • Standard flask clamps model can be used with clamps sold separately
Heavy duty, low profile, compact footprint Orbital Shakers available in 3 different configurations provide the flexibility for using a mix of different size and shape vessels on a single platform with shaking speed up to 300 RPM.
Accessories information: Accessory Universal Ratcheting Clamp is for use with the Ratcheting Mini-300 Orbital-Genie.
Accessory blue adhering mat is an alternative to flask clamps or roller bars for securing vessels to the Mini-300 Orbital-Genie platform. It is ideal for containers such as bottles and dishes that cannot be held with conventional clamps or roller bars. Patented material is naturally adhering. Rinse clean to renew adhering properties. Can only be used with SI-M16xx or SI-M17xx.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty