Would you like to log in again?
ADAPTOR ELER E1140 ROT ANODE SS FOR ANLY
Supplier: EBERBACH CORP
Electrode Adaptor Rotating Anode Stainless Steel^ Electrode Adaptor, Rotating Anode, Stainless Steel, electrodes are held in the holder by tightening a collar with a small screw driver, For Electro-Analyzer E1000 apparatus for standard determinations of metals by electro-deposition process
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
ADAPTOR ELER E1140 ROT ANODE SS FOR ANLY 75836-264 E1050.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty