Would you like to log in again?
HOMOGENIZER CON-TORQUE TSU 230 V W/ STND
Supplier: EBERBACH CORP
Con-Torque Tissue Homogenizer 230V^ Con-Torque Tissue Homogenizer, Stirrer Speed: 0 - 400 rpm, Maximum Torque: 196 oz-in, Electrical: 230V at 50/60Hz, with power unit, chucking device and universal support, with integral clamp fits on support rods, Dimension (without stand): 6.5 in x 6 in x 14 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
HOMOGENIZER CON-TORQUE TSU 230 V W/ STND 75836-268 E2355.25 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty