Would you like to log in again?
SHAKER HV DTY RECIPROCAL 230V 43X22X10IN
Supplier: EBERBACH CORP
Reciprocating Shaker Heavy-Duty 43 x 22 x 10in^ Reciprocating Shaker Heavy-Duty, Speed: 2-Speed 160 - 240 osc/min (+/- 5%), Stroke: 1.5 in, Maximum Capacity: 250 lbs, Electrical: 230V at 50/60hz, designed to shake large containers or large quantities of smaller containers, Dimensions: 43 x 22 x 10in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER HV DTY RECIPROCAL 230V 43X22X10IN 75836-292 E5902.HD.25 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty