Would you like to log in again?
SHAKER HV DTY RECIPROCAL DIG LARGE 115 V
Supplier: EBERBACH CORP
Digital Reciprocal Shaker Heavy Duty 39 x 20 x 11 in^ Digital Reciprocal Shaker Heavy Duty, Speed: 60 - 240 rpm, Stroke: 1.5 in, Maximum Capacity: 100 lbs, Chassis Material: Powder Coat Steel or Stainless Steel, Electrical: 115V at 50/60hz, with BLDC speed control, Dimensions: 39 x 20 x 11 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER HV DTY RECIPROCAL DIG LARGE 115 V 75836-296 E5903.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty