Would you like to log in again?
SHAKER MID RNG RECIPROCAL DIG 115V-50 HZ
Supplier: EBERBACH CORP
Digital Reciprocal Shaker Mid Range 115V^ Digital Reciprocal Shaker Mid Range, Speed: 60 - 260 osc/min, Stroke: 1.5 in, Maximum Capacity: 50 lbs, Controls: Digital, Electrical: 115V at 50/60hz, Blue Powder Coat finish, Variable-Speed, with digital speed control, Dimension: 24.5 x 13.5 x 10 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER MID RNG RECIPROCAL DIG 115V-50 HZ 75836-322 E6003.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty