Would you like to log in again?
SHAKER FIXED SPD RECIPROCAL 230 V BNCHTP
Supplier: EBERBACH CORP
Benchtop Reciprocal Shaker Fixed Speed 230V^ Benchtop Reciprocal Shaker Fixed Speed, Stroke: 1.5 in, Maximum Capacity: 40 lbs, Speed: 180 or 280 osc/min, Controls: Physical Switch (High/Low Speed), Electrical: 230V at 50/60hz, Designed for general purpose shaking needs, Dimension: 16.5 x 11 x 10 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER FIXED SPD RECIPROCAL 230 V BNCHTP 75836-326 E6010.25 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty