Would you like to log in again?
SHAKER DIG RECIPROCAL 115V-50HZ VRBL SPD
Supplier: EBERBACH CORP
Benchtop Reciprocal Shaker Digital Variable Speed 115V^ Benchtop Reciprocal Shaker Digital Variable Speed, Stroke: 1.5in, Speed: 60-260 osc/min, Capacity: 40 lbs, Controls: Digital, Electrical: 115V, Power Unit. Blue Powder Coat finish, designed for heavy-duty shaking needs, Dimension: 16.5x11x10 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER DIG RECIPROCAL 115V-50HZ VRBL SPD 75836-330 E6013.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty