Would you like to log in again?
TUMBLER PROGRAMMABLE INVERSION 115V-50HZ
Supplier: EBERBACH CORP
Inversion Tumbler Programmable 0 - 100 rpm^ Inversion Tumbler Programmable, Rotating Speed: 0 - 100 rpm, Rotation Direction: Forward and Reverse, Maximum Load Capacity: 12.5 lbs, Container Capacity: 6 Jars/Bottles up to 7in tall and 3in in diameter, Electrical: 115V, Dimension: 37 x12 x15 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUMBLER PROGRAMMABLE INVERSION 115V-50HZ 75836-400 EL655.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty