Would you like to log in again?
TUMBLER HIGH CAP PRGMBL INVERSION 115 V
Supplier: EBERBACH CORP
Programmable Inversion Tumbler High Capacity 48 x16.25x19 in^ Programmable Inversion Tumbler High Capacity, designed to mix wet or dry samples, Rotating Speed: 0 - 100 rpm, Rotation Direction: Forward and Reverse, Maximum Load Capacity: 25 lbs, Electrical: 115V at 50/60hz, Dimension: 48x16.25x19 in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
TUMBLER HIGH CAP PRGMBL INVERSION 115 V 75836-450 EL660.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty