Would you like to log in again?
SHAKER QUECHERS HND MOTN 230V 33X12X10IN
Supplier: EBERBACH CORP
QuEChERS Hand Motion Shaker 100-600 osc/min^ QuEChERS Hand Motion Shaker, Shaking Speed: 100-600osc/min, Stroke: 10.5 Degrees, Max Load Capacity: 10 lbs, for extracting, dissolving and emulsifying samples, Electrical: 230V at 50/60hz, includes a digital timer, rubber feet, Dimension: 33 inx12inx10in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER QUECHERS HND MOTN 230V 33X12X10IN 75836-458 EL680.Q.25 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty