Would you like to log in again?
SHAKER HMDTY RSTNT INCBR READY ORB 115 V
Supplier: EBERBACH CORP
Incubator Ready Orbital Shaker 30 - 320 rpm^ Incubator Ready Orbital Shaker, Speed: 30 - 320 rpm, Orbit: 1 in, Maximum Capacity: 70 lbs, CO2 Rating: 8%, Humidity Rating: 100%, Temperature Rating: 32 - 140 F, Electrical: 115V at 50/60hz, Blue Powder Coat finish, Dimensions(Power unit): 23x21.3x8in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER HMDTY RSTNT INCBR READY ORB 115 V 75836-466 E6140.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty