Would you like to log in again?
SHAKER LARGE ORBITAL 115V-50HZ 30-320RPM
Supplier: EBERBACH CORP
Orbital Shaker Large 34 x 27 in^ Orbital Shaker Large, Speed: 30 - 320 rpm, Orbit: Standard: 1 in - Optional: 0.5, 2, or 3 in, Mounting: Floor, Suction cup feet or optional Floor Stand, Electrical: 115V at 50/60hz, IP67 Wash Down. Data output via Ethernet, Profibus, Dimensions(Power unit): 34 x 27in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHAKER LARGE ORBITAL 115V-50HZ 30-320RPM 75836-468 E6145.00 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty