Would you like to log in again?
M121 Scepter™ Non-sterile, Lightweight, Cut Resistant Liner, Wells Lamont
Supplier: Wells Lamont
Scepter™
M121XL M121S M121M M121L M121XS
75836-512BX 115.92 USD InStock
75836-512 75836-520 75836-518 75836-514 75836-516
M121 Scepter™ Non-sterile, Lightweight, Cut Resistant Liner, Wells Lamont
Gloves
These lightweight and cut-resistant liners are manufactured from polyester and stainless steel.
  • Thin gauge stainless steel wrapped in white polyester
  • Launderable 15 times without shrinking
  • Moderate tactile sensitivity
  • Latex-free
  • Ambidextrous
  • Packaged 10 liners per box
These non-sterile gloves can be used as a glove or liner.
Certifications: ANSI Level 4, A4
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty