Would you like to log in again?
SteriPlast® Scoops for Foodstuffs, PS, Bürkle
Supplier: Bürkle
SteriPlast® SteriPlast
5378-5019 5378-3015 5378-3019 5378-3020 5378-5015 5378-3001 5378-3007 5378-3005 5378-3013 5378-3003
75836-620PK 98.17 USD InStock
75836-620 75848-664 75836-566 76501-362 76501-372 75836-562 76501-396 75836-564 75836-618 75836-616
SteriPlast® Scoops for Foodstuffs, PS, Bürkle
Scoops
The SteriPlast® food scoop is ideal for powders, granulates and pastes.
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Individually packaged and sterilized by gamma rays
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.
Certifications: Production, assembly and packaging according to cleanroom class 7. Complies with EU food requirements and FDA regulations.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty