Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Transglutaminase Activity Assay Kit (Colorimetric), BioVision

Supplier: BioVision
AB204700-100TEST
75836-744EA 1342.87 USD InStock
75836-744
Transglutaminase Activity Assay Kit (Colorimetric), BioVision
Assays Protein Assays
Validated using many sample types.

  • Simple, rapid & convenient
  • The assay can detect transglutaminase activity ~10 µU or 80 ng of recombinant hTG2 enzyme
  • Detection method- Absorbance (525 nm)
  • Applications- Measurement of Transglutaminase activity

Transglutaminases (EC 2.3.2.13) are calcium dependent enzymes that catalyze the post-translational modification of proteins by formation of isopeptide bonds. This occurs either through protein cross-linking via formation of γ-glutamyl-ε-lysine bonds or through incorporation of primary amines at selected peptide-bound glutamine residues. The transglutaminase enzyme family comprises the intracellular forms (TG1, TG3 and TG5) expressed mostly in the epithelial tissue; TG2 which is both intracellular and extracellular and expressed in various tissue types; TG4 which is expressed in the prostate gland; factor XIII which is expressed in blood; TG6 and TG7, whose tissue distribution is unknown and band 4.2 (lacking enzymatic activity) which is present on erythrocyte membranes. Transglutaminases also exhibit GTPase, phosphodiesterase and protein kinase activity. Transglutaminases are associated with certain neurological and autoimmune disorders and also cancer. BioVision’s Transglutaminase Activity Assay kit utilizes the deamidation reaction of the transglutaminase enzyme with a donor and acceptor substrate resulting in the formation of a hydroxamate product. The hydroxamate product reacts with the Stop Solution forming a purple complex that can be measured colorimetrically at 525 nm. The limit of quantification of this assay is ~10 µU or 80 ng of recombinant hTG2 enzyme.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty