Would you like to log in again?
Cytochrome P450 2D6 (CYP2D6) Activity Assay Kit (Fluorometric), BioVision
Supplier: BioVision
AB211078-200TEST
75836-752EA 873.64 USD InStock
75836-752
Cytochrome P450 2D6 (CYP2D6) Activity Assay Kit (Fluorometric), BioVision
Assays Protein Assays
Highly Sensitive; Includes human CYP2D6 - NADPH Reductase as positive control.
  • Simple, convenient, highly sensitive
  • Fluorescent assay enables easy determination of CYP2D6 activity in a variety of biological samples
  • Kit includes CYP2D6 inhibitor Quinidine and a stable, recombinant human CYP2D6 co-expressed with NADPH Reductase as a positive control
  • The substrate shows low background and a high signal-to-noise ratio
Cytochrome P450 2D6 (CYP2D6, EC 1.14.14.1) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. CYPs are membrane-bound hemeproteins responsible for Phase I biotransformation reactions, in which lipophilic drugs and other xenobiotic compounds are converted to more hydrophilic products to facilitate excretion from the body. CYP2D6 catalyzes oxidation of lipophilic bases with an aromatic ring and a nitrogen atom and is highly expressed in liver and brain tissue. The enzyme is responsible for metabolism of nearly 25% of all small molecule drugs commonly used by humans, particularly psychiatric drugs such as antidepressants, antipsychotics and stimulants. The CYP2D6 gene is highly polymorphic in the human population, with CYP2D6 activity ranging from complete metabolic deficiency to ultra-rapid metabolism. Due to this wide phenotypic variability, CYP2D6 is frequently implicated in drug toxicity and clinical drug/drug interactions. In addition, for drugs whose pharmacological activity requires metabolism from a pro-drug form, CYP2D6 inhibition or allelic deficiency can lead to decreased drug efficacy. BioVision’s CYP2D6 Activity Assay Kit enables rapid measurement of native or recombinant CYP2D6 activity in biological samples such as liver microsomes. The assay utilizes a non-fluorescent CYP2D6-selective substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 390/468 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. CYP2D6 specific activity is calculated by running parallel reactions in the presence and absence of the potent CYP2D6-selective inhibitor quinidine and subtracting any residual activity detected with the inhibitor present. The kit contains a complete set of reagents sufficient for performing 100 sets of paired reactions (in the presence and absence of inhibitor).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty