Would you like to log in again?
EZLabel™ Protein FITC Labeling Kit, BioVision
Supplier: BioVision
AB288089-5TEST
75836-762EA 345.5 USD InStock
75836-762
EZLabel™ Protein FITC Labeling Kit, BioVision
Protein Linking and Labeling Systems
Easy to use Kit to label upto 5mg of Protein wz FITC for multiple applications such as ELISA,Wblot.
  • All necessary reagents included.
  • Fast and efficient labeling of proteins with FITC
  • Easy to use Kit to label upto 5 mg of Protein with FITC for multiple applications such as ELISA, western blot, Immunohistochemistry, Immunofluorescence and FACS analysis
BioVision’s EZLabel™ Protein-FITC Labeling Kit provides an easy way to label proteins with Fluorescein Isothiocyanate (FITC) in a user-friendly spin column format. FITC is an ideal dye for fluorescent labeling of proteins among the most commonly used fluorescent dyes for labeling proteins. Each spin column provided in the kit can be used to label up to 1 mg of the target protein. The kit provides all of the reagents necessary to perform five labeling reactions using up to 1 mg of protein per reaction. FITC-labeled protein has an excitation and emission wavelengths at 494 nm and 520 nm respectively, and can be directly used for downstream applications including ELISA, western blot, Immunohistochemistry, Immunofluorescence, and FACS analysis, etc.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty