Would you like to log in again?
Cytochrome P450 2C9 (CYP2C9) Inhibitor Screening Kit (Fluorometric), BioVision
Supplier: BioVision
AB284519-100TEST
75837-074EA 719.04 USD InStock
75837-074
Cytochrome P450 2C9 (CYP2C9) Inhibitor Screening Kit (Fluorometric), BioVision
Assays Protein Assays
High-throughput compatible,quick screening for CYP2C9 inhibitors or ligands; Includes human CYP2C9
  • Simple, highly sensitive, high-throughput compatible
  • Rapid screening of CYP2C9 inhibitors or ligands
  • Kit includes the CYP2C9 inhibitor Sulfaphenazole and a stable, recombinant human CYP2C9 co-expressed with NADPH Reductase
Cytochrome P450 2C9 (CYP2C9, EC 1.14.14.1) is a member of the cytochrome P450 monooxidase (CYP) family of microsomal xenobiotic metabolism enzymes. CYPs are membrane-bound hemeproteins responsible for Phase I biotransformation reactions, in which lipophilic drugs and other xenobiotic compounds are converted to more hydrophilic products to facilitate excretion from the body. CYP2C9 is primarily expressed in liver and catalyzes oxidation of small, weakly acidic or hydrophobic molecules containing an aromatic moiety and at least one hydrogen bond donor. Isoforms of the CYP2C subfamily are responsible for metabolism of nearly 20% of all small molecule drugs commonly used by humans. The human CYP2C9 gene is highly polymorphic and CYP2C9 single nucleotide polymorphisms (SNPs) have been implicated in clinical drug/drug interactions involving widely-prescribed drugs with narrow therapeutic indices such as warfarin, phenytoin and indomethacin. In addition, for drugs whose pharmacological activity requires metabolism from a pro-drug form (such as the antihypertensive drug losartan), CYP2C9 inhibition or allelic deficiency can lead to decreased drug efficacy. BioVision’s CYP2C9 Inhibitor Screening Kit enables rapid screening of drugs and other new chemical entities (NCEs) for compound-CYP2C9 interaction in a reliable, high-throughput fluorescence-based assay. The kit provides a yeast microsomal preparation of human CYP2C9 and human cytochrome P450 reductase (CPR) enzymes. The assay utilizes a non-fluorescent CYP2C9 substrate that is converted into a highly fluorescent metabolite detected in the visible range (Ex/Em = 415/502 nm), ensuring a high signal-to-background ratio with little interference by autofluorescence. The kit contains a complete set of reagents sufficient for performing 100 reactions in a 96-well plate format.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty